2-Bromo-2’,4’-difluoropropiophenone

Supplier: Apollo Scientific
Danger

PC502325-1G PC502325-250MG PC502325-5G
APOSPC502325-1GEA 321 EUR
APOSPC502325-1G APOSPC502325-250MG APOSPC502325-5G
2-Bromo-2’,4’-difluoropropiophenone
2-Bromo-2’,4’-difluoropropiophenone
Formula: C₉H₇BrF₂O
MW: 249,052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07847871
CAS Number: 135206-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR