3-Fluoro-5-iodo-4-methoxybenzyl alcohol 95%

Supplier: Apollo Scientific
Warning

PC502373-1G PC502373-250MG
APOSPC502373-1GEA 1760 EUR
APOSPC502373-1G APOSPC502373-250MG
3-Fluoro-5-iodo-4-methoxybenzyl alcohol 95%
3-Fluoro-5-iodo-4-methoxybenzyl alcohol
Formula: C₈H₈FIO₂
MW: 282,051 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30830103
CAS Number: 2090747-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR