3-Bromo-2,5,6-trifluorobenzaldehyde 97%
Supplier: Apollo Scientific
PC502420-10G
PC502420-1G
PC502420-5G
PC502420-25G
PC502420-100G
APOSPC502420-10GEA
1040
EUR
APOSPC502420-10G
APOSPC502420-1G
APOSPC502420-5G
APOSPC502420-25G
APOSPC502420-100G
3-Bromo-2,5,6-trifluorobenzaldehyde 97%
3-Bromo-2,5,6-trifluorobenzaldehyde
Formula:
C₇H₂BrF₃O MW: 238,989 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26687662 CAS Number: 137234-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...