3-Bromo-2,5,6-trifluoroaniline
Supplier: Apollo Scientific
|
Warning
|
PC502421-1G
PC502421-5G
APOSPC502421-1GEA
654
EUR
APOSPC502421-1G
APOSPC502421-5G
3-Bromo-2,5,6-trifluoroaniline
3-Bromo-2,5,6-trifluoroaniline
Formula:
C₆H₃BrF₃N MW: 225,994 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22627830 CAS Number: 1269440-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...