4-Bromo-2,3-difluoroiodobenzene 97%

Supplier: Apollo Scientific
Warning

Synonyms: 1-Bromo-2,3-difluoro-4-iodobenzene

PC50251-1G PC50251-250MG PC50251-5G
APOSPC50251-1GEA 259 EUR
APOSPC50251-1G APOSPC50251-250MG APOSPC50251-5G
4-Bromo-2,3-difluoroiodobenzene 97%
4-Bromo-2,3-difluoroiodobenzene
Formula: C₆H₂BrF₂I
MW: 318,885 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878200
CAS Number: 207989-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR