4-Iodo-2-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
PC502640-1G
PC502640-5G
APOSPC502640-1GEA
65.5
EUR
APOSPC502640-1G
APOSPC502640-5G
4-Iodo-2-(trifluoromethyl)benzoic acid
4-Iodo-2-(trifluoromethyl)benzoic acid
Formula:
C₈H₄F₃IO₂ MW: 316,016 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09881756 CAS Number: 954815-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...