Methyl 3-nitro-5-(trifluoromethyl)benzoate

Supplier: Apollo Scientific

PC502652-1G PC502652-5G
APOSPC502652-1GEA 58.2 EUR
APOSPC502652-1G APOSPC502652-5G
Methyl 3-nitro-5-(trifluoromethyl)benzoate
Methyl 3-nitro-5-(trifluoromethyl)benzoate
Formula: C₉H₆F₃NO₄
MW: 249,143 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06637356
CAS Number: 22227-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR