3-Bromo-4-(trifluoromethyl)benzonitrile 98%

Supplier: Apollo Scientific

PC502704-1G PC502704-5G PC502704-10G
APOSPC502704-1GEA 94.4 EUR
APOSPC502704-1G APOSPC502704-5G APOSPC502704-10G
3-Bromo-4-(trifluoromethyl)benzonitrile 98%
3-Bromo-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃BrF₃N
MW: 250,015 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185387
CAS Number: 1212021-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR