5-Bromo-2-nitrobenzyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC502887-1G PC502887-250MG
APOSPC502887-1GEA 609 EUR
APOSPC502887-1G APOSPC502887-250MG
5-Bromo-2-nitrobenzyl fluoride 95%
5-Bromo-2-nitrobenzyl fluoride
Formula: C₇H₅BrFNO₂
MW: 234,023 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31634832
CAS Number: 2244083-48-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR