4-Bromo-2-methylbenzyl fluoride 95%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Bromo-1-(fluoromethyl)-2-methylbenzene

PC502898-1G PC502898-250MG PC502898-5G
APOSPC502898-1GEA 318 EUR
APOSPC502898-1G APOSPC502898-250MG APOSPC502898-5G
4-Bromo-2-methylbenzyl fluoride 95%
4-Bromo-2-methylbenzyl fluoride
Formula: C₈H₈BrF
MW: 203,052 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22492378
CAS Number: 1824142-81-5
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR