2-Bromo-3-(trifluoromethyl)benzyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC502904-1G PC502904-250MG
APOSPC502904-1GEA 609 EUR
APOSPC502904-1G APOSPC502904-250MG
2-Bromo-3-(trifluoromethyl)benzyl fluoride 95%
Formula: C₈H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD31634838
CAS Number: 2244087-97-4
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR