2-Bromo-5-fluorophenylisocyanate
Supplier: Apollo Scientific
|
Danger
|
PC503001-1G
PC503001-25G
PC503001-5G
APOSPC503001-1GEA
21.9
EUR
APOSPC503001-1G
APOSPC503001-25G
APOSPC503001-5G
2-Bromo-5-fluorophenylisocyanate
2-Bromo-5-fluorophenylisocyanate
May form small amount of sediment
Formula:
C₇H₃BrFNO Storage Temperature: Refrigerator |
MDL Number:
MFCD09945449 CAS Number: 1016819-26-3 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...