3-(Trifluoromethoxy)phenylisocyanate

Supplier: Apollo Scientific
Danger

PC503004-1G PC503004-25G PC503004-5G
APOSPC503004-1GEA 87 EUR
APOSPC503004-1G APOSPC503004-25G APOSPC503004-5G
3-(Trifluoromethoxy)phenylisocyanate
3-(Trifluoromethoxy)phenylisocyanate

May form small amount of sediment

Formula: C₈H₄F₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16877177
CAS Number: 55225-86-0
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR