Ethyl 5-fluorobenzofuran-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
PC50379-1G
PC50379-250MG
APOSPC50379-1GEA
498
EUR
APOSPC50379-1G
APOSPC50379-250MG
Ethyl 5-fluorobenzofuran-2-carboxylate
Ethyl 5-fluorobenzofuran-2-carboxylate
Formula:
C₁₁H₉FO₃ MW: 208,186 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09038171 CAS Number: 93849-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...