1-(3-Bromoprop-1-ynyl)-2-fluoro-benzene 95%
Supplier: Apollo Scientific
|
Warning
|
PC50401-1G
PC50401-250MG
PC50401-5G
APOSPC50401-1GEA
78.3
EUR
APOSPC50401-1G
APOSPC50401-250MG
APOSPC50401-5G
1-(3-Bromoprop-1-ynyl)-2-fluoro-benzene 95%
1-(3-Bromoprop-1-ynyl)-2-fluoro-benzene
Formula:
C₉H₆BrF Storage Temperature: Freezer |
MDL Number:
MFCD22482443 CAS Number: 1020153-63-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...