4-Chloro-3-(trifluoromethoxy)bromobenzene 97%

Supplier: Apollo Scientific

Synonyms: 4-Bromo-1-chloro-2-(trifluoromethoxy)benzene, 5-Bromo-2-chloro-1-trifluoromethoxybenzene

PC50436-10G PC50436-1G PC50436-5G PC50436-25G
APOSPC50436-10GEA 575 EUR
APOSPC50436-10G APOSPC50436-1G APOSPC50436-5G APOSPC50436-25G
4-Chloro-3-(trifluoromethoxy)bromobenzene 97%
4-Chloro-3-(trifluoromethoxy)bromobenzene
Formula: C₇H₃BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD09878140
CAS Number: 406232-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR