4-Bromo-2,3,6-trifluorobenzonitrile 95%
Supplier: Apollo Scientific
PC50547-1G
PC50547-250MG
PC50547-5G
APOSPC50547-1GEA
574
EUR
APOSPC50547-1G
APOSPC50547-250MG
APOSPC50547-5G
4-Bromo-2,3,6-trifluorobenzonitrile 95%
4-Bromo-2,3,6-trifluorobenzonitrile
Formula:
C₇HBrF₃N MW: 235,989 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32669843 CAS Number: 2514941-93-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...