4-Ethynyl-2-fluorobenzonitrile 95%

Supplier: Apollo Scientific

PC50563-1G PC50563-250MG
APOSPC50563-1GEA 609 EUR
APOSPC50563-1G APOSPC50563-250MG
4-Ethynyl-2-fluorobenzonitrile 95%
4-Ethynyl-2-fluorobenzonitrile
Formula: C₉H₄FN
MW: 145,133 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11618086
CAS Number: 167858-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR