6-Bromo-2,3,4-trifluorophenol
Supplier: Apollo Scientific
PC50634-1G
PC50634-250MG
APOSPC50634-1GEA
524
EUR
APOSPC50634-1G
APOSPC50634-250MG
6-Bromo-2,3,4-trifluorophenol
6-Bromo-2,3,4-trifluorophenol
Formula:
C₆H₂BrF₃O MW: 226,979 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22414729 CAS Number: 1517643-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...