8-Bromo-6-fluoroquinolin-4(1H)-one

Supplier: Apollo Scientific
Warning

PC508105-1G PC508105-250MG
APOSPC508105-1GEA 551 EUR
APOSPC508105-1G APOSPC508105-250MG
8-Bromo-6-fluoroquinolin-4(1H)-one
8-Bromo-6-fluoroquinolin-4(1H)-one
Formula: C₉H₅BrFNO
MW: 242,045 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12192827
CAS Number: 1019016-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR