6-Bromo-8-(trifluoromethyl)quinolin-4(1H)-one
Supplier: Apollo Scientific
|
Warning
|
PC508110-1G
PC508110-250MG
APOSPC508110-1GEA
241
EUR
APOSPC508110-1G
APOSPC508110-250MG
6-Bromo-8-(trifluoromethyl)quinolin-4(1H)-one
6-Bromo-8-(trifluoromethyl)quinolin-4(1H)-one
Formula:
C₁₀H₅BrF₃NO MW: 292,052 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31745734 CAS Number: 1065092-55-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...