8-Chloro-5-fluoroquinolin-4(1H)-one

Supplier: Apollo Scientific
Warning

PC508116-1G PC508116-250MG
APOSPC508116-1GEA 241 EUR
APOSPC508116-1G APOSPC508116-250MG
8-Chloro-5-fluoroquinolin-4(1H)-one
8-Chloro-5-fluoroquinolin-4(1H)-one
Formula: C₉H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD31745738
CAS Number: 1065092-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR