8-Chloro-5-(trifluoromethyl)quinolin-4(1H)-one

Supplier: Apollo Scientific
Warning

PC508117-1G PC508117-250MG
APOSPC508117-1GEA 241 EUR
APOSPC508117-1G APOSPC508117-250MG
8-Chloro-5-(trifluoromethyl)quinolin-4(1H)-one
8-Chloro-5-(trifluoromethyl)quinolin-4(1H)-one
Formula: C₁₀H₅ClF₃NO
MW: 247,601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31745739
CAS Number: 1065092-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR