2-(4-Amino-2,6-difluorophenyl)propanoic acid

Supplier: Apollo Scientific

PC510017-1G PC510017-5G
APOSPC510017-1GEA 499 EUR
APOSPC510017-1G APOSPC510017-5G
2-(4-Amino-2,6-difluorophenyl)propanoic acid
2-(4-Amino-2,6-difluorophenyl)propanoic acid
Formula: C₉H₉F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD16495906
CAS Number: 1226776-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR