6-[3,5-Bis(trifluoromethyl)phenyl]picolinic acid

Supplier: Apollo Scientific

PC510059-1G PC510059-5G
APOSPC510059-1GEA 499 EUR
APOSPC510059-1G APOSPC510059-5G
6-[3,5-Bis(trifluoromethyl)phenyl]picolinic acid
6-[3,5-Bis(trifluoromethyl)phenyl]picolinic acid
Formula: C₁₄H₇F₆NO₂
MW: 335,201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410088
CAS Number: 887982-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR