6-Bromo-3-chloro-5-fluoro-1H-indazole

Supplier: Apollo Scientific
Warning

PC51021-100MG PC51021-1G PC51021-500MG
APOSPC51021-100MGEA 79.9 EUR
APOSPC51021-100MG APOSPC51021-1G APOSPC51021-500MG
6-Bromo-3-chloro-5-fluoro-1H-indazole
6-Bromo-3-chloro-5-fluoro-1H-indazole
Formula: C₇H₃BrClFN₂
Storage Temperature: Ambient
MDL Number: MFCD18384873
CAS Number: 1286734-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR