Methyl 5-Fluoro-6-methylnicotinate
Supplier: Apollo Scientific
PC510214-1G
PC510214-250MG
PC510214-5G
APOSPC510214-1GEA
672
EUR
APOSPC510214-1G
APOSPC510214-250MG
APOSPC510214-5G
Methyl 5-Fluoro-6-methylnicotinate
Methyl 5-Fluoro-6-methylnicotinate
Formula:
C₈H₈FNO₂ MW: 169,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18803129 CAS Number: 1253383-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...