4-Fluoro-7-methylindole
Supplier: Apollo Scientific
PC510216-1G
PC510216-250MG
PC510216-5G
APOSPC510216-1GEA
499
EUR
APOSPC510216-1G
APOSPC510216-250MG
APOSPC510216-5G
4-Fluoro-7-methylindole
4-Fluoro-7-methylindole
Formula:
C₉H₈FN MW: 149,165 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09027019 CAS Number: 313337-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...