6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole
Supplier: Apollo Scientific
|
Warning
|
PC51022-1G
PC51022-250MG
APOSPC51022-1GEA
611
EUR
APOSPC51022-1G
APOSPC51022-250MG
6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole
6-Bromo-5-fluoro-3-iodo-1-methyl-1H-indazole
Formula:
C₈H₅BrFIN₂ MW: 354,946 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18384874 CAS Number: 1286734-96-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...