2-Amino-6-bromo-5-fluoro-3-methylbenzoic acid
Supplier: Apollo Scientific
PC510264-1G
PC510264-250MG
APOSPC510264-1GEA
916
EUR
APOSPC510264-1G
APOSPC510264-250MG
2-Amino-6-bromo-5-fluoro-3-methylbenzoic acid
2-Amino-6-bromo-5-fluoro-3-methylbenzoic acid
Formula:
C₈H₇BrFNO₂ MW: 248,049 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28977408 CAS Number: 1694614-97-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...