4-Chloro-5,7-difluoroquinazoline
Supplier: Apollo Scientific
PC510282-1G
PC510282-250MG
APOSPC510282-1GEA
1840
EUR
APOSPC510282-1G
APOSPC510282-250MG
4-Chloro-5,7-difluoroquinazoline
4-Chloro-5,7-difluoroquinazoline
Formula:
C₈H₃ClF₂N₂ MW: 200,573 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18377822 CAS Number: 791602-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...