Methyl 2-(4-fluoro-1H-indol-3-yl)acetate
Supplier: Apollo Scientific
PC510350-250MG
PC510350-1G
APOSPC510350-250MGEA
256
EUR
APOSPC510350-250MG
APOSPC510350-1G
Methyl 2-(4-fluoro-1H-indol-3-yl)acetate
Methyl 2-(4-fluoro-1H-indol-3-yl)acetate
Formula:
C₁₁H₁₀FNO₂ MW: 207,201 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24617138 CAS Number: 5159-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...