Methyl 3-amino-4,4,4-trifluorobutanoate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Methyl 3-amino-4,4,4-trifluorobutyrate
PC51059-1G
PC51059-250MG
APOSPC51059-1GEA
350
EUR
APOSPC51059-1G
APOSPC51059-250MG
Methyl 3-amino-4,4,4-trifluorobutanoate
Methyl 3-amino-4,4,4-trifluorobutanoate
Formula:
C₅H₈F₃NO₂ MW: 171,118 g/mol Boiling Pt: 50 °C (1 mm Hg) Density: 1,293 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD09265033 CAS Number: 748746-28-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...