5-Bromo-2-fluoro-3-hydroxypyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-3-hydroxy-5-bromopyridine, 5-Bromo-2-fluoropyridin-3-ol

PC51114-5G
APOSPC51114-5GEA 94.4 EUR
APOSPC51114-5G
5-Bromo-2-fluoro-3-hydroxypyridine 98%
5-Bromo-2-fluoro-3-hydroxypyridine
Formula: C₅H₃BrFNO
MW: 191,986 g/mol
Melting Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD11040265
CAS Number: 1012084-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR