5-Bromo-2-chloro-4-fluorophenylacetonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(5-Bromo-2-chloro-4-fluorophenyl)acetonitrile, 5-Bromo-2-chloro-4-fluorobenzyl cyanide
PC51137-25G
APOSPC51137-25GEA
148
EUR
APOSPC51137-25G
5-Bromo-2-chloro-4-fluorophenylacetonitrile 97%
5-Bromo-2-chloro-4-fluorophenylacetonitrile
Formula:
C₈H₄BrClFN MW: 248,479 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD23166759 CAS Number: 1426290-08-5 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...