Methyl 2,2-difluoropropanoate

Supplier: Apollo Scientific
Danger

Synonyms: Methyl 2,2-difluoropropionate

PC51164-1G PC51164-5G
APOSPC51164-1GEA 101 EUR
APOSPC51164-1G APOSPC51164-5G
Methyl 2,2-difluoropropanoate
Methyl 2,2-difluoropropanoate
Formula: C₄H₆F₂O₂
MW: 124,086 g/mol
Boiling Pt: 93…95 °C
Density: 1,182 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD20489374
CAS Number: 38650-84-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR