4-[(Trifluoromethyl)sulphonyl]phenol 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Hydroxyphenyl trifluoromethyl sulphone
PC51178-1G
PC51178-250MG
APOSPC51178-1GEA
261
EUR
APOSPC51178-1G
APOSPC51178-250MG
4-[(Trifluoromethyl)sulphonyl]phenol 97%
4-[(Trifluoromethyl)sulphonyl]phenol
Formula:
C₇H₅F₃O₃S MW: 226,173 g/mol Melting Pt: 119…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD22053286 CAS Number: 432-84-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...