2,6-Dibromo-4-fluoro-1,3-benzothiazole 99%
Supplier: Apollo Scientific
|
Warning
|
PC51328-1G
PC51328-250MG
APOSPC51328-1GEA
264
EUR
APOSPC51328-1G
APOSPC51328-250MG
2,6-Dibromo-4-fluoro-1,3-benzothiazole 99%
2,6-Dibromo-4-fluoro-1,3-benzothiazole
Formula:
C₇H₂Br₂FNS MW: 310,969 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09748620 CAS Number: 1188226-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...