4-Fluoro-1H-pyrrolo[2,3-b]pyridin-5-ol 97%

Supplier: Apollo Scientific
Warning

PC51345-1G PC51345-250MG
APOSPC51345-1GEA 469 EUR
APOSPC51345-1G APOSPC51345-250MG
4-Fluoro-1H-pyrrolo[2,3-b]pyridin-5-ol 97%
4-Fluoro-1H-pyrrolo[2,3-b]pyridin-5-ol
Formula: C₇H₅FN₂O
MW: 152,126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11656012
CAS Number: 651744-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR