5-Bromo-4-fluoro-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Apollo Scientific
Warning

PC51346-1G PC51346-250MG
APOSPC51346-1GEA 532 EUR
APOSPC51346-1G APOSPC51346-250MG
5-Bromo-4-fluoro-1H-pyrrolo[2,3-b]pyridine 98%
5-Bromo-4-fluoro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₄BrFN₂
MW: 215,022 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12031517
CAS Number: 1172067-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR