3-Hydroxy-5-(trifluoromethyl)benzyl alcohol
Supplier: Apollo Scientific
|
Warning
|
PC51525-1G
PC51525-250MG
APOSPC51525-1GEA
415
EUR
APOSPC51525-1G
APOSPC51525-250MG
3-Hydroxy-5-(trifluoromethyl)benzyl alcohol
3-Hydroxy-5-(trifluoromethyl)benzyl alcohol
Formula:
C₈H₇F₃O₂ MW: 192,135 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18397635 CAS Number: 1261611-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...