Methyl 2,6-difluoro-4-(trifluoromethyl)benzoate

Supplier: Apollo Scientific
Warning

PC52035-1G PC52035-250MG
APOSPC52035-1GEA 437 EUR
APOSPC52035-1G APOSPC52035-250MG
Methyl 2,6-difluoro-4-(trifluoromethyl)benzoate
Methyl 2,6-difluoro-4-(trifluoromethyl)benzoate
Formula: C₉H₅F₅O₂
MW: 240,127 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28897189
CAS Number: 1980049-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR