1-Cyclopropyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylic acid 97%
Supplier: Apollo Scientific
Synonyms:
1-Cyclopropyl-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
PC520515-100G
PC520515-25G
APOSPC520515-100GEA
698
EUR
APOSPC520515-100G
APOSPC520515-25G
1-Cyclopropyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylic acid 97%
1-Cyclopropyl-6,7,8-trifluoro-4-oxo-quinoline-3-carboxylic acid
Formula:
C₁₃H₈F₃NO₃ MW: 283,203 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01173812 CAS Number: 94695-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...