Butyl 2,2-difluorocyclopropanecarboxylate 95%
Supplier: Apollo Scientific
|
Warning
|
PC520549-1G
PC520549-250MG
PC520549-5G
APOSPC520549-1GEA
46.4
EUR
APOSPC520549-1G
APOSPC520549-250MG
APOSPC520549-5G
Butyl 2,2-difluorocyclopropanecarboxylate 95%
Butyl 2,2-difluorocyclopropanecarboxylate
Formula:
C₈H₁₂F₂O₂ MW: 178,176 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06797747 CAS Number: 260352-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...