2-Fluoro-5-iodo-4-methylaniline, tech

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-5-fluoro-2-iodotoluene, 2-Fluoro-5-iodo-p-toluidine

PC52061-1G PC52061-250MG PC52061-5G
APOSPC52061-1GEA 200 EUR
APOSPC52061-1G APOSPC52061-250MG APOSPC52061-5G
2-Fluoro-5-iodo-4-methylaniline, tech
2-Fluoro-5-iodo-4-methylaniline
Formula: C₇H₇FIN
MW: 251,04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27992342
CAS Number: 1820711-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR