2,6-Dichloro-5-fluoro-3-pyridinecarboxaldehyde 97%
Supplier: Apollo Scientific
Synonyms:
2,6-Dichloro-5-fluoronicotinaldehyde
PC520715-10G
PC520715-1G
PC520715-5G
APOSPC520715-10GEA
1630
EUR
APOSPC520715-10G
APOSPC520715-1G
APOSPC520715-5G
2,6-Dichloro-5-fluoro-3-pyridinecarboxaldehyde 97%
2,6-Dichloro-5-fluoro-3-pyridinecarboxaldehyde
Formula:
C₆H₂Cl₂FNO MW: 193,991 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15528788 CAS Number: 1260758-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...