3-Chloro-4-methyl-benzenesulphonyl fluoride 95%
Supplier: Apollo Scientific
PC520762-1G
PC520762-5G
APOSPC520762-1GEA
187
EUR
APOSPC520762-1G
APOSPC520762-5G
3-Chloro-4-methyl-benzenesulphonyl fluoride 95%
3-Chloro-4-methyl-benzenesulphonyl fluoride
Formula:
C₇H₆ClFO₂S MW: 208,638 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22192509 CAS Number: 25300-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...