3-(Difluoromethoxy)-2-fluoro-pyridine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(Difluoromethoxy)-2-fluoropyridine

PC520799-1G PC520799-5G PC520799-25G
APOSPC520799-1GEA 26.1 EUR
APOSPC520799-1G APOSPC520799-5G APOSPC520799-25G
3-(Difluoromethoxy)-2-fluoro-pyridine 98%
3-(Difluoromethoxy)-2-fluoro-pyridine
Formula: C₆H₄F₃NO
Storage Temperature: Ambient
MDL Number: MFCD13185836
CAS Number: 1214377-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR