6-(Trifluoromethylsulphanyl)pyridin-3-amine 95%

Supplier: Apollo Scientific

PC520817-1G PC520817-250MG PC520817-500MG
APOSPC520817-1GEA 1060 EUR
APOSPC520817-1G APOSPC520817-250MG APOSPC520817-500MG
6-(Trifluoromethylsulphanyl)pyridin-3-amine 95%
6-(Trifluoromethylsulphanyl)pyridin-3-amine
Formula: C₆H₅F₃N₂S
MW: 194,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12094672
CAS Number: 1153767-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR