3-(Trifluoromethyl)cyclobutanecarboxylic acid 95%
Supplier: Apollo Scientific
|
Warning
|
PC520819-250MG
PC520819-500MG
APOSPC520819-250MGEA
768
EUR
APOSPC520819-250MG
APOSPC520819-500MG
3-(Trifluoromethyl)cyclobutanecarboxylic acid 95%
3-(Trifluoromethyl)cyclobutanecarboxylic acid
Formula:
C₆H₇F₃O₂ MW: 168,114 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15526994 CAS Number: 1093750-93-6 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...